missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (3S)-3-amino-3-phenylpropanoate, 97%

Product Code. 10745293 Shop All Thermo Scientific Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10745293

Brand: Thermo Scientific Acros 337892500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120686-18-2 | C13H20NO2 | 222.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120686-18-2
Molecular Formula C13H20NO2
Molecular Weight (g/mol) 222.31
MDL Number MFCD00798310
InChI Key TYYCBAISLMKLMT-NSHDSACASA-O
PubChem CID 7021093
SMILES CC(C)(C)OC(=O)C[C@H]([NH3+])C1=CC=CC=C1

Specifications

Color Colorless to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Refractive Index 1.4960 to 1.498
Specific Rotation -22
Synonym s-tert-butyl 3-amino-3-phenylpropanoate, tert-butyl 3s-3-amino-3-phenylpropanoate, benzenepropanoic acid, b-amino-, 1,1-dimethylethyl ester, bs, s-tert-butyl3-amino-3-phenylpropanoate, t-butyl 3s-3-amino-3-phenylpropanoate, s-alpha-2-oxo-2-tert-butoxyethyl benzyl amine, tert-butyl-3s-3-amino-3-phenylpropanoate, benzenepropanoic acid,b-amino-,1,1-dimethylethyl ester, bs
IUPAC Name tert-butyl (3S)-3-amino-3-phenylpropanoate
Specific Rotation Condition - 22.00 (20.00°C c=1,CHCl3)
Quantity 250 mg
Formula Weight 221.3
Percent Purity 97%
Physical Form Slightly Viscous Liquid
Chemical Name or Material tert-Butyl (3S)-3-amino-3-phenylpropanoate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butyl (3S)-3-amino-3-phenylpropanoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.