Learn More
tert-Butyl 4-(2-(aminomethyl)phenyl)piperazine-1-carboxylate, ≥97%, Thermo Scientific™
£72.50 - £215.00
Chemical Identifiers
CAS | 174855-53-9 |
---|---|
Molecular Formula | C16H25N3O2 |
Molecular Weight (g/mol) | 291.395 |
MDL Number | MFCD04115066 |
InChI Key | UINNZXQKTNAOBL-UHFFFAOYSA-N |
Synonym | tert-butyl 4-2-aminomethyl phenyl piperazine-1-carboxylate, 1-boc-4-2-aminomethyl phenyl piperazine, 1-boc-4-2-aminomethyl-phenyl-piperazine, 1-2-aminomethyl-phenyl-piperazine-4-carboxylic acid tert-butyl ester, 1-piperazinecarboxylicacid, 4-2-aminomethyl phenyl-, 1,1-dimethylethyl ester, 1-2-aminomethyl-phenyl-piperazine-4-carboxylic a, 2-4-t-butoxycarbonylpiperazin-1-yl phenylmethylamine, 4-2-aminomethyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-aminomethyl phenyl-1-piperazinecarboxylic acid tert-butyl esterShow More |
PubChem CID | 4961274 |
IUPAC Name | tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10085822
|
Thermo Scientific Maybridge
CC30713CB |
250 mg | Amber Glass Bottle |
£72.50
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10117722
|
Thermo Scientific Maybridge
CC30713DA |
1 g | Amber Glass Bottle |
£215.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
174855-53-9 | |
291.395 | |
UINNZXQKTNAOBL-UHFFFAOYSA-N | |
4961274 | |
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2CN |
C16H25N3O2 | |
MFCD04115066 | |
tert-butyl 4-2-aminomethyl phenyl piperazine-1-carboxylate, 1-boc-4-2-aminomethyl phenyl piperazine, 1-boc-4-2-aminomethyl-phenyl-piperazine, 1-2-aminomethyl-phenyl-piperazine-4-carboxylic acid tert-butyl ester, 1-piperazinecarboxylicacid, 4-2-aminomethyl phenyl-, 1,1-dimethylethyl ester, 1-2-aminomethyl-phenyl-piperazine-4-carboxylic a, 2-4-t-butoxycarbonylpiperazin-1-yl phenylmethylamine, 4-2-aminomethyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-aminomethyl phenyl-1-piperazinecarboxylic acid tert-butyl esterShow More | |
tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate |
Specifications
174855-53-9 | |
100.0 | |
C16H25N3O2 | |
tert-butyl 4-2-aminomethyl phenyl piperazine-1-carboxylate, 1-boc-4-2-aminomethyl phenyl piperazine, 1-boc-4-2-aminomethyl-phenyl-piperazine, 1-2-aminomethyl-phenyl-piperazine-4-carboxylic acid tert-butyl ester, 1-piperazinecarboxylicacid, 4-2-aminomethyl phenyl-, 1,1-dimethylethyl ester, 1-2-aminomethyl-phenyl-piperazine-4-carboxylic a, 2-4-t-butoxycarbonylpiperazin-1-yl phenylmethylamine, 4-2-aminomethyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-aminomethyl phenyl-1-piperazinecarboxylic acid tert-butyl ester | |
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2CN | |
291.395 | |
291.39 | |
Tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate |
96.5 | |
97% | |
MFCD04115066 | |
UINNZXQKTNAOBL-UHFFFAOYSA-N | |
tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate | |
4961274 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
Your input is important to us. Please complete this form to provide feedback related to the content on this product.