missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl 4-(2-aminophenyl)tetrahydro-1(2H)-pyrazinecarboxylate, 97%, Thermo Scientific™ Product Code.: 10259202

tert-Butyl 4-(2-aminophenyl)tetrahydro-1(2H)-pyrazinecarboxylate, 97%, Thermo Scientific™

Product Code. 10259202
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10259202

Brand: Thermo Scientific Maybridge CC30714CB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 170017-74-0
Molecular Formula C15H23N3O2
Molecular Weight (g/mol) 277.37
MDL Number MFCD04115046
InChI Key LNDQGWAWYKFYAO-UHFFFAOYSA-N
Synonym 1-boc-4-2-aminophenyl piperazine, tert-butyl 4-2-aminophenyl piperazine-1-carboxylate, 2-4-boc-piperazin-1-yl aniline, tert-butyl 4-2-aminophenyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-2-aminophenyl-piperazine, 4-2-amino-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 2-4-tert-butoxycarbonyl piperazin-1-yl aniline, 4-2-aminophenyl piperazine, n1-boc protected, pubchem12189, acmc-1bzzw
PubChem CID 2795530
IUPAC Name tert-butyl 4-(2-aminophenyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=CC=C1N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 277.37
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-(2-aminophenyl)tetrahydro-1(2H)-pyrazinecarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl 4-(2-aminophenyl)tetrahydro-1(2H)-pyrazinecarboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.