missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-[2-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate, 97%, Thermo Scientific™

Product Code. p-4813673
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10533314

Brand: Thermo Scientific Maybridge CC30709DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 179250-28-3
Molecular Formula C16H24N2O3
Molecular Weight (g/mol) 292.379
InChI Key MQZUQGHXDLMAKT-UHFFFAOYSA-N
Synonym 2-4-n-boc-piperazinyl benzyl alcohol, tert-butyl 4-2-hydroxymethyl phenyl tetrahydro-1 2h-pyrazinecarboxylate, tert-butyl 4-2-hydroxymethyl phenyl piperazine-1-carboxylate, pubchem11335, 1-boc-4-hydroxymethyl-phenyl-piperazine, 1-boc-4-2-hydroxymethyl-phenyl-piperazine, 1-2-hydroxymethyl-phenyl-piperazine-4-carboxylic, 1-t-butoxycarbonyl-4-2-hydroxymethyl-phenyl-piperazine, 4-2-hydroxymethylphenyl piperazine-1-carboxylic acid t-butyl ester
PubChem CID 7127802
IUPAC Name tert-butyl 4-[2-(hydroxymethyl)phenyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2CO

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 292.38
Chemical Name or Material tert-Butyl 4-[2-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.