missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(3-aminobenzyl)piperazine-1-carboxylate, 97%, Thermo Scientific™

Product Code. 10780364
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 361345-40-6
Molecular Formula C16H25N3O2
Molecular Weight (g/mol) 291.395
InChI Key VKSINZNBWVJILC-UHFFFAOYSA-N
Synonym tert-butyl 4-3-aminobenzyl piperazine-1-carboxylate, 1-boc-4-3-aminobenzyl piperazine, 3-4-boc-piperazin-1-yl-methyl aniline, tert-butyl 4-3-aminophenyl methyl piperazine-1-carboxylate, 3-1-boc-piperazin-4-yl-methyl-aniline, tert-butyl 4-3-aminophenyl methyl piperazinecarboxylate, 4-3-amino-benzyl-piperazine-1-carboxylic acid tert-butyl ester, pubchem12209, acmc-1ah36, 4-3-aminobenzyl piperazine, n1-boc protected
PubChem CID 7127799
IUPAC Name tert-butyl 4-[(3-aminophenyl)methyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC(=CC=C2)N

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 291.39
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-(3-aminobenzyl)piperazine-1-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'