missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate, 97%, Maybridge™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 311.23 |
Percent Purity | 97% |
Chemical Name or Material | tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate, 97%, Maybridge™ >
Spot an opportunity for improvement?Share a Content Correction