missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Maybridge tert-Butyl 4-[4-(aminomethyl)pyrid-2-yl]piperazine-1-carboxylate, 1g, 97%

Brand:  Thermo Scientific Maybridge CC50613DA

tert-Butyl 4-[4-(aminomethyl)pyrid-2-yl]piperazine-1-carboxylate, 1g, 97%

Code : 31

Additional Details : CAS Number : 910036-87-2 Weight : 0.05100kg Transport : UN number : 3259 Chem class : 8 Pack group : III

Product Code. 10205574

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Description

Description

TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers