missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(4-formyl-1,3-thiazol-2-yl)piperidine-1-carboxylate, ≥97%, Thermo Scientific™

Product Code. 10748823
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 869901-02-0
Molecular Formula C14H20N2O3S
Molecular Weight (g/mol) 296.385
MDL Number MFCD06658981
InChI Key XLGKMJFDRZHAEV-UHFFFAOYSA-N
PubChem CID 7172298
IUPAC Name tert-butyl 4-(4-formyl-1,3-thiazol-2-yl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C2=NC(=CS2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym tert-butyl 4-4-formylthiazol-2-yl piperidine-1-carboxylate, tert-butyl 4-4-formyl-1,3-thiazol-2-yl piperidine-1-carboxylate, tert-butyl 4-4-formyl-1,3-thiazol-2-yl tetrahydro-1 2h-pyridinecarboxylate, tert-butyl 4-4-formylthiazol-2-yl piperidine-1-ca, tert-butyl4-4-formylthiazol-2-yl piperidine-1-carboxylate, 1,1-dimethylethyl 4-4-formyl-2-thiazolyl-1-piperidinecarboxylate, 4-4-formyl-thiazol-2-yl-piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylicacid, 4-4-formyl-2-thiazolyl-, 1,1-dimethylethyl ester, 4-4-formyl-2-thiazolyl-1-piperidinecarboxylic acid,1,1-dimethylethyl ester
Formula Weight 296.39
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-(4-formyl-1, 3-thiazol-2-yl)tetrahydro-1(2H)-pyridinecarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.