missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 4-[5-(hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine-1-carboxylate, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 312.43 |
Percent Purity | 95% |
Chemical Name or Material | Tert-butyl 4-[5-(hydroxymethyl)-4-methyl-1, 3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction