missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(aminocarbonyl)tetrahydropyridine-1(2H)-carboxylate, 97%, Thermo Scientific™

Product Code. 11376965
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 91419-48-6
Molecular Formula C11H20N2O3
Molecular Weight (g/mol) 228.292
MDL Number MFCD02180953
InChI Key YHFUWPUJUMZXBD-UHFFFAOYSA-N
Synonym 1-n-boc-piperidine-4-carboxamide, 1-n-boc-isonipecotamide, 1-n-boc-4-piperidinecarboxamide, 1-tert-butoxycarbonyl piperidine-4-carboxamide, n-boc-4-piperidine carboxamide, 1-boc-4-piperidinecarboxamide, 4-carbamoyl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-aminocarbonyl tetrahydropyridine-1 2h-carboxylate, 1-piperidinecarboxylic acid, 4-aminocarbonyl-, 1,1-dimethylethyl ester, tert-butyl 4-carbamoylpiperidinecarboxylate
PubChem CID 2735646
IUPAC Name tert-butyl 4-carbamoylpiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C(=O)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 228.29
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-(aminocarbonyl)tetrahydropyridine-1(2H)-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'