missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl 4-formyl-3,5-dimethyl-1H-pyrazole-1-carboxylate, 97%, Thermo Scientific™ Product Code.: 10413214

tert-Butyl 4-formyl-3,5-dimethyl-1H-pyrazole-1-carboxylate, 97%, Thermo Scientific™

Product Code. 10413214
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 10413214

Brand: Thermo Scientific Maybridge CC31904DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 844891-13-0
Molecular Formula C11H16N2O3
Molecular Weight (g/mol) 224.26
MDL Number MFCD06659076
InChI Key AZJWODKWFMBMCM-UHFFFAOYSA-N
PubChem CID 2795543
IUPAC Name tert-butyl 4-formyl-3,5-dimethyl-1H-pyrazole-1-carboxylate
SMILES CC1=NN(C(=O)OC(C)(C)C)C(C)=C1C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Synonym tert-butyl 4-formyl-3,5-dimethyl-1h-pyrazole-1-carboxylate, tert-butyl4-formyl-3,5-dimethyl-1h-pyrazole-1-carboxylate, tert-butyl 4-formyl-3,5-dimethylpyrazolecarboxylate, 3,5-dimethyl-4-formyl-1h-pyrazole, n1-boc protected, 1h-pyrazole-1-carboxylicacid, 4-formyl-3,5-dimethyl-, 1,1-dimethylethyl ester
Formula Weight 224.26
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-formyl-3, 5-dimethyl-1H-pyrazole-1-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl 4-formyl-3,5-dimethyl-1H-pyrazole-1-carboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.