missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl acetoacetate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
tert-Butyl acetoacetate is used as intermediate for the syntheses of different organic chemicals. Raw material in pharmaceutical Industry, In agrochemical industry. Reagent for Acyloin synthesis and ?,β - unsaturated ketones synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyl acetoacetate is used as intermediate for the syntheses of different organic chemicals. Raw material in pharmaceutical Industry, In agrochemical industry. Reagent for Acyloin synthesis and ∞,β - unsaturated ketones synthesis.
Solubility
Soluble in water (9g/L).
Notes
Keep container tightly closed. Store away from oxidizing agents.
tert-Butyl acetoacetate is used as intermediate for the syntheses of different organic chemicals. Raw material in pharmaceutical Industry, In agrochemical industry. Reagent for Acyloin synthesis and ∞,β - unsaturated ketones synthesis.
Solubility
Soluble in water (9g/L).
Notes
Keep container tightly closed. Store away from oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -38°C |
Density | 0.961 |
pH | 5.7 |
Boiling Point | 71°C to 72°C (11 mmHg) |
Flash Point | 76°C (169°F) |
Odor | Pleasant |
Refractive Index | 1.42 |
Quantity | 100 g |
Beilstein | 1680303 |
Solubility Information | Soluble in water (9g/L). |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'