missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals Product Code.: 11324827

tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 11324827
250 mL
Click to view available options
Quantity:
50 mL
250 mL
1 L
4 x 1 L
This item is not returnable. View return policy

Product Code. 11324827

Brand: Thermo Scientific Alfa Aesar 041857.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

tert-Butyl alcohol is used as a solvent and an ethanol denaturant. It is an active ingredient in paint remover. It acts as an intermediate in the production of methyl tertiary butyl ether and ethyl tertiary butyl ether by reaction with methanol and ethanol. It is also used in the preparation of tert-butyl chloride, tert-butyl hydroperoxide, isobutylene and methyl methacrylate. It serves as a fuel additive as well as a gasoline octane booster.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004464
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcoholShow MoreShow Less
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Melting Point 25°C
Color Colorless
Density 0.786 g/mL
Boiling Point 83°C
Flash Point 11°C (52°F)
Assay Percent Range 99.8%
Packaging ChemSeal™ bottles
Linear Formula (CH3)3COH
Refractive Index 1.387
Quantity 250 mL
UN Number UN1120
Beilstein 906698
Merck Index 14,1542
Solubility Information Miscible with water,ethanol,ethyl ether and chloroform.
Formula Weight 74.12
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material tert-Butyl alcohol, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ15Y932 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog Number041857.M1,041857.AD,041857.AK,041857.K2
Lot NumberJ30P703 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number041857.M1,041857.AD,041857.AK,041857.K2
Lot NumberG28N754 Certificate TypeCertificate of Analysis Date25/01/2023 Catalog Number041857.M1,041857.AD,041857.AK,041857.K2
Lot NumberR05H723 Certificate TypeCertificate of Analysis Date04/11/2022 Catalog Number041857.M1,041857.AD,041857.AK,041857.K2
Lot NumberT30H711 Certificate TypeCertificate of Analysis Date13/06/2022 Catalog Number041857.M1,041857.AD,041857.AK,041857.K2
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl alcohol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : tert-Butyl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.