missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Alcohol, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

£207.00 - £653.00

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15518524
View Documents
Avantor J.T.Baker
9056-01
500 mL HDPE Pressure Bottom Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15528524
View Documents
Avantor J.T.Baker
9056-05
4 L Poly Bottle
£653.00
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

75-65-0
74.123
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
CHEBI:45895
CC(C)(C)O
C4H10O
DKGAVHZHDRPRBM-UHFFFAOYSA-N
6386
2-methylpropan-2-ol

Specifications

75-65-0
C4H10O
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
CC(C)(C)O
74.123
CHEBI:45895
BAKER ANALYZED™ ACS Reagent
11.00°C
1L = 0.78kg
DKGAVHZHDRPRBM-UHFFFAOYSA-N
2-methylpropan-2-ol
6386
74.12
tert-Butyl Alcohol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • tert-Butyl Alcohol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

SDS
Documents

Documents

Product Certifications