missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tert-Butyl Alcohol, Certified AR for Analysis, Fisher Chemical™
£81.20 - £199.00
Chemical Identifiers
CAS | 75-65-0 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.12 |
MDL Number | 4464 |
InChI Key | DKGAVHZHDRPRBM-UHFFFAOYSA-N |
IUPAC Name | 2-methylpropan-2-ol |
SMILES | CC(C)(C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10181750
|
Fisher Chemical
B/5251/08 |
500 mL |
£81.20
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10233900
|
Fisher Chemical
B/5251/17 |
2.5 L |
£199.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
75-65-0 | |
74.12 | |
DKGAVHZHDRPRBM-UHFFFAOYSA-N | |
CC(C)(C)O |
C4H10O | |
4464 | |
2-methylpropan-2-ol |
Specifications
7 | |
75-65-0 | |
HDPE Bottle | |
C4H10O | |
36mbar at 20°C | |
DKGAVHZHDRPRBM-UHFFFAOYSA-N | |
2-methylpropan-2-ol | |
74.12g/mol |
25°C | |
83°C | |
Liquid | |
4464 | |
6.43 mPaS (25°C) | |
CC(C)(C)O | |
74.12 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- tert-Butanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.