missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl bromide, 98+%, stab. with potassium carbonate, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
Description
tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.
Solubility
Immiscible with water.
Notes
Incompatible with strong bases and strong oxidizing agents.
tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.
Solubility
Immiscible with water.
Notes
Incompatible with strong bases and strong oxidizing agents.
Specifications
CAS | 507-19-7 |
Melting Point | °C to 20°C |
Density | 1.210 |
Boiling Point | 74°C to 76°C |
Flash Point | 18°C |
Assay Percent Range | 98%, stabilized with potassium carbonate |
Molecular Formula | C4H9Br |
Refractive Index | 1.428 |
MDL Number | MFCD00000125 |
Quantity | 25 g |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'