missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl bromoacetate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.
Solubility
Miscible with ethanol, chloroform and ethyl acetate. Immiscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.
Solubility
Miscible with ethanol, chloroform and ethyl acetate. Immiscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.33 |
Boiling Point | 175°C |
Flash Point | 49°C (120°F) |
Odor | Pungent |
Linear Formula | BrCH2CO2C(CH3)3 |
Refractive Index | 1.445 |
Quantity | 50 g |
UN Number | UN2924 |
Beilstein | 1753010 |
Solubility Information | Miscible with ethanol,chloroform and ethyl acetate. Immiscible with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'