Learn More
tert-Butyl bromoacetate, 99%, Thermo Scientific Chemicals
£23.00 - £284.00
Chemical Identifiers
CAS | 5292-43-3 |
---|---|
Molecular Formula | C6H11BrO2 |
Molecular Weight (g/mol) | 195.06 |
MDL Number | MFCD00000188 |
InChI Key | BNWCETAHAJSBFG-UHFFFAOYSA-N |
Synonym | tert-butyl bromoacetate, t-butyl bromoacetate, acetic acid, bromo-, 1,1-dimethylethyl ester, bromoacetic acid tert-butyl ester, tert-butylbromoacetate, acetic acid, bromo-, tert-butyl ester, brch2c o oc ch3 3, acetic acid, 2-bromo-, 1,1-dimethylethyl ester, bromo-acetic acid tert-butyl ester, tert-butylbromacetat |
PubChem CID | 79177 |
IUPAC Name | tert-butyl 2-bromoacetate |
SMILES | CC(C)(C)OC(=O)CBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10764831
|
Thermo Scientific Acros
154340100 |
10 mL | Glass bottle |
£23.00
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10665012
|
Thermo Scientific Acros
154340500 |
50 mL | Glass bottle |
£84.70
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10540902
|
Thermo Scientific Acros
154342500 |
250 mL | Glass bottle |
£284.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5292-43-3 | |
195.06 | |
BNWCETAHAJSBFG-UHFFFAOYSA-N | |
79177 | |
CC(C)(C)OC(=O)CBr |
C6H11BrO2 | |
MFCD00000188 | |
tert-butyl bromoacetate, t-butyl bromoacetate, acetic acid, bromo-, 1,1-dimethylethyl ester, bromoacetic acid tert-butyl ester, tert-butylbromoacetate, acetic acid, bromo-, tert-butyl ester, brch2c o oc ch3 3, acetic acid, 2-bromo-, 1,1-dimethylethyl ester, bromo-acetic acid tert-butyl ester, tert-butylbromacetat | |
tert-butyl 2-bromoacetate |
Specifications
5292-43-3 | |
1.3380g/mL | |
49°C | |
98.5% min. (GC) | |
BrCH2CO2C(CH3)3 | |
MFCD00000188 | |
1.338 | |
Solubility in water: immiscible | |
CC(C)(C)OC(=O)CBr | |
195.06 | |
195.06 | |
Liquid |
Colorless to Yellow | |
50°C (10.0 mmHg) | |
Authentic | |
C6H11BrO2 | |
1.4440 to 1.446 | |
02, I, 96 | |
tert-butyl bromoacetate, t-butyl bromoacetate, acetic acid, bromo-, 1,1-dimethylethyl ester, bromoacetic acid tert-butyl ester, tert-butylbromoacetate, acetic acid, bromo-, tert-butyl ester, brch2c o oc ch3 3, acetic acid, 2-bromo-, 1,1-dimethylethyl ester, bromo-acetic acid tert-butyl ester, tert-butylbromacetat | |
BNWCETAHAJSBFG-UHFFFAOYSA-N | |
tert-butyl 2-bromoacetate | |
79177 | |
99% | |
tert-Butyl bromoacetate, 99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 226-133-6
Recommended Storage : Refrigerator +4°C
RUO – Research Use Only