Learn More
tert-Butyl chloroacetate, 97%, Thermo Scientific Chemicals
£21.00 - £154.00
Chemical Identifiers
CAS | 107-59-5 |
---|---|
Molecular Formula | C6H11ClO2 |
Molecular Weight (g/mol) | 150 |
MDL Number | MFCD00000930 |
InChI Key | KUYMVWXKHQSIAS-UHFFFAOYSA-N |
Synonym | tert-butyl chloroacetate, chloroacetic acid tert-butyl ester, acetic acid, chloro-, 1,1-dimethylethyl ester, t-butyl chloroacetate, tert-butylchloroacetate, acetic acid, chloro-, tert-butyl ester, chloroacetic acid t-butyl ester, acetic acid, 2-chloro-, 1,1-dimethylethyl ester, t-butylchloroacetate, t-butyl-chloroacetate |
PubChem CID | 66052 |
IUPAC Name | tert-butyl 2-chloroacetate |
SMILES | CC(C)(C)OC(=O)CCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10356451
|
Thermo Scientific Acros
347230250 |
25 g | Glass bottle |
£21.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10254751
|
Thermo Scientific Acros
347231000 |
100 g | Glass bottle |
£46.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10753524
|
Thermo Scientific Acros
347235000 |
500 g | Glass bottle |
£154.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-59-5 | |
150 | |
KUYMVWXKHQSIAS-UHFFFAOYSA-N | |
66052 | |
CC(C)(C)OC(=O)CCl |
C6H11ClO2 | |
MFCD00000930 | |
tert-butyl chloroacetate, chloroacetic acid tert-butyl ester, acetic acid, chloro-, 1,1-dimethylethyl ester, t-butyl chloroacetate, tert-butylchloroacetate, acetic acid, chloro-, tert-butyl ester, chloroacetic acid t-butyl ester, acetic acid, 2-chloro-, 1,1-dimethylethyl ester, t-butylchloroacetate, t-butyl-chloroacetate | |
tert-butyl 2-chloroacetate |
Specifications
107-59-5 | |
1.0530g/mL | |
46°C | |
96% min. (GC) | |
ClCH2CO2C(CH3)3 | |
MFCD00000930 | |
01,86; 03,36 | |
1.053 | |
Solubility in water: <0.1g/L (20°C). Other solubilities: soluble in isopropanol,1,2-dichloroethane,toluene | |
CC(C)(C)OC(=O)CCl | |
150 | |
150 | |
Liquid |
Colorless to Yellow | |
48.0°C to 49.0°C (11.0 mmHg) | |
Authentic | |
C6H11ClO2 | |
1.4220 to 1.4240 | |
02, III, 444 | |
15, 1564 | |
tert-butyl chloroacetate, chloroacetic acid tert-butyl ester, acetic acid, chloro-, 1,1-dimethylethyl ester, t-butyl chloroacetate, tert-butylchloroacetate, acetic acid, chloro-, tert-butyl ester, chloroacetic acid t-butyl ester, acetic acid, 2-chloro-, 1,1-dimethylethyl ester, t-butylchloroacetate, t-butyl-chloroacetate | |
KUYMVWXKHQSIAS-UHFFFAOYSA-N | |
tert-butyl 2-chloroacetate | |
66052 | |
97% | |
tert-Butyl chloroacetate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Very toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
Harmful if swallowed.
Toxic if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-506-1
- tert-Butyl chloroacetate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only