missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl disulfide, 98%, Thermo Scientific Chemicals

Product Code. 11935731
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-06-5
Molecular Formula C8H18S2
Molecular Weight (g/mol) 178.35
MDL Number MFCD00008838
InChI Key BKCNDTDWDGQHSD-UHFFFAOYSA-N
Synonym di-tert-butyl disulfide, disulfide, bis 1,1-dimethylethyl, 1,2-di-tert-butyldisulfane, di-tert-butyl disulphide, tert-butyl disulfide, t-butyl disulfide, 2-tert-butyldisulfanyl-2-methylpropane, di-t-butyl disulfide, t-butyldisulfide, 2,2'-dithiobis 2-methylpropane
PubChem CID 66060
IUPAC Name 2-(tert-butyldisulfanyl)-2-methylpropane
SMILES CC(C)(C)SSC(C)(C)C

Specifications

Melting Point -5.0°C
Color Yellow
Density 0.9300g/mL
Boiling Point 198.0°C to 204.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)3C]2S2
Refractive Index 1.4882 to 1.4902
Quantity 2.5 kg
Beilstein 01,IV,1638
Specific Gravity 0.93
Solubility Information Solubility in water: insoluble
Formula Weight 178.35
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material tert-Butyl disulfide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl disulfide
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'