missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals

£15.88 - £399.99

Chemical Identifiers

CAS 75-91-2, 7732-18-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
InChI Key CIHOLLKRGTVIJN-UHFFFAOYSA-N
Synonym tert-butyl hydroperoxide, tbhp, t-butyl hydroperoxide, tert-butylhydroperoxide, perbutyl h, t-butylhydroperoxide, cadox tbh, hydroperoxide, 1,1-dimethylethyl, 1,1-dimethylethyl hydroperoxide, terc. butylhydroperoxid
PubChem CID 6410
ChEBI CHEBI:64090
IUPAC Name 2-hydroperoxy-2-methylpropane
SMILES CC(C)(C)OO
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10703571
View Documents Promotion Details
Thermo Scientific Acros
180340050
5 g Plastic bottle
£15.88
5g
Please to purchase this item. Need a web account? Register with us today!
10266090
View Documents Promotion Details
Thermo Scientific Acros
180342500
250 g Plastic bottle
£24.92
250g
Please to purchase this item. Need a web account? Register with us today!
10347360
View Documents Promotion Details
Thermo Scientific Acros
180345000
500 g Plastic bottle
£37.19
500g
Please to purchase this item. Need a web account? Register with us today!
10649103
View Documents Promotion Details
Thermo Scientific Acros
180340025
2.5 kg Plastic bottle
£125.63
2.50kg
Please to purchase this item. Need a web account? Register with us today!
11914761
View Documents Promotion Details
Thermo Scientific Acros
180340100
10 kg Plastic Can
£399.99
10kg
Please to purchase this item. Need a web account? Register with us today!
Currently Unavailable: We have no information when this product will be available
View alternative products
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-91-2, 7732-18-5
90.12
tert-butyl hydroperoxide, tbhp, t-butyl hydroperoxide, tert-butylhydroperoxide, perbutyl h, t-butylhydroperoxide, cadox tbh, hydroperoxide, 1,1-dimethylethyl, 1,1-dimethylethyl hydroperoxide, terc. butylhydroperoxid
CHEBI:64090
CC(C)(C)OO
C4H10O2
CIHOLLKRGTVIJN-UHFFFAOYSA-N
6410
2-hydroperoxy-2-methylpropane

Specifications

75-91-2, 7732-18-5
Colorless
37.0°C (15.0 mmHg)
C4H10O2
1.3860 to 1.3880
01,88; 02,49; 03,37; 05,75; 06,81; 07,43; 08,62; 09,78; 10,64; 11,91; 12,88; 14,60; 15,55; 16,53; 17,56
0.94
Solubility in water: miscible. Other solubilities: soluble in hydroxide solutions,soluble in most common organic solvents
CC(C)(C)OO
90.12
CHEBI:64090
90.12
tert-Butyl hydroperoxide
-2.8°C
0.9400g/mL
43°C
(CH3)3COOH
01,IV,1616
15,1573
tert-butyl hydroperoxide, tbhp, t-butyl hydroperoxide, tert-butylhydroperoxide, perbutyl h, t-butylhydroperoxide, cadox tbh, hydroperoxide, 1,1-dimethylethyl, 1,1-dimethylethyl hydroperoxide, terc. butylhydroperoxid
CIHOLLKRGTVIJN-UHFFFAOYSA-N
2-hydroperoxy-2-methylpropane
6410
4.1 mPa.s (20°C)
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 200-915-7

Product Identifier
  • tert-Butyl hydroperoxide, 70% solution in water

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 2
  • Organic peroxide Type E & F
  • Skin sensitiser Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
  • H242-Heating may cause a fire.
  • H226-Flammable liquid and vapour.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only