missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl isothiocyanate, 97%, Thermo Scientific Chemicals

Product Code. 11347785
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

tert-Butyl isothiocyanate is a tool for the modification of the pore sizes on the top layer of heterostructured surface mounted metal-organic frameworks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl isothiocyanate is a tool for the modification of the pore sizes on the top layer of heterostructured surface mounted metal-organic frameworks.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Incompatible with amines, alcohols and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-42-1
Molecular Formula C5H9NS
Molecular Weight (g/mol) 115.19
MDL Number MFCD00004816
InChI Key ZFWFRTVIIMTOLY-UHFFFAOYSA-N
Synonym tert-butyl isothiocyanate, t-butyl isothiocyanate, propane, 2-isothiocyanato-2-methyl, tert-butylisothiocyanate, tert.-butyl isothiocyanate, 2-isothiocyanato-2-methyl-propane, tert butyl isothiocyanate, tert-butyliso thiocyanate, butyl isothiocyanate, tert, t-butylisothiocyanate
PubChem CID 11544
IUPAC Name 2-isothiocyanato-2-methylpropane
SMILES CC(C)(C)N=C=S

Specifications

Melting Point 10°C to 12°C
Density 0.908
Boiling Point 141°C to 145°C
Flash Point 38°C (100°F)
Odor Pungent
Refractive Index 1.481
Quantity 5 g
UN Number UN2929
Beilstein 1361854
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 115.2
Percent Purity 97%
Chemical Name or Material tert-Butyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.