missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals Product Code.: 10501422

tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals

Product Code. 10501422
25 L, Metal Drum
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic Drum
This item is not returnable. View return policy

Product Code. 10501422

Brand: Thermo Scientific Acros 177040250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropaneShow MoreShow Less
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -110.0°C
Color White to Yellow
Density 0.7404g/mL
Boiling Point 54.0°C to 56.0°C
Quantity 25 L
Flash Point −28°C
Infrared Spectrum Authentic
Packaging Metal Drum
Physical Form Powder or Needles
Refractive Index 1.3675 to 1.3695
Assay Percent Range 99%
Specific Gravity 0.7404
Linear Formula (CH3)3COCH3
Viscosity 0.36 mPa.s (20°C)
Beilstein 01, 381
Water 0.01% Max. (Coulometric)
Fieser 04,333
Merck Index 15, 6106
Solubility Information Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g
Formula Weight 88.15
Percent Purity 99%
Grade Pure
Chemical Name or Material tert-Butyl methyl ether, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464996 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number177040025,177042500,177040010,177040100,17704024,177040250
Lot NumberA0459084 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog Number177040025,177042500,177040010,177040100,17704024,177040250
Lot NumberA0454615 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number177040025,177042500,177040010,177040100,17704024,177040250
Lot NumberA0450714 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number177040025,177042500,177040010,177040100,17704024,177040250
Lot NumberA0395681 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number177040025,177042500,177040010,177040100,177040250
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.