missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl methyl ether, HPLC Grade, 99+%
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
Packungsgröße:
1 L
16 L
4 L
Beschreibung
Used as a Chromatography solvent. Acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline(oxygenates).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Melting Point | -109°C |
| Density | 0.742 g/mL |
| Boiling Point | 55°C to 56°C |
| Quantity | 1 L |
| Flash Point | -25°C (-13°F) |
| Odor | Mild |
| Physical Form | Liquid |
| Refractive Index | 1.369 |
| UN Number | UN2350 |
| Beilstein | 1730942 |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- tert-Butyl methyl ether
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- MIXTURE LIST-Contains : tert-Butyl methyl ether
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur