missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-{2-[2-amino-4-(trifluoromethyl)anilino]ethyl}carbamate, 95%, Thermo Scientific™

Catalog Number 10502282
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 215655-42-8
CAS Min % 94.5
CAS Max % 95.0
Assay Percent Range 95%
Molecular Formula C14H20F3N3O2
Linear Formula CF3C6H3(NH2)NH(CH2)2NHC(O)OC(CH3)3
MDL Number MFCD00728760
Quantity 1 g
Synonym tert-butyl n-2-2-amino-4-trifluoromethyl anilino ethyl carbamate, tert-butyl n-2-2-amino-4-trifluoromethyl phenyl amino ethyl carbamate, tert-butyl 2-2-amino-4-trifluoromethyl phenyl amino ethylcarbamate, tert-butyl-n-2-2-amino-4-trifluoromethyl anilino ethyl carbamate, n-2-2-amino-4-trifluoromethyl phenyl amino ethyl tert-butoxy carboxamide, maybridge1_000225, tert-butyl 2-2-amino-4-trifluoromethyl anilino ethyl carbamate, t-butyl n-2-2-amino-4-trifluoromethyl anilino ethylcarbamate
InChI Key LBPALYCQELHDIC-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NCCNC1=C(C=C(C=C1)C(F)(F)F)N
IUPAC Name tert-butyl N-[2-[2-amino-4-(trifluoromethyl)anilino]ethyl]carbamate
Molecular Weight (g/mol) 319.328
PubChem CID 2773437
Formula Weight 319.33
Percent Purity 95%
Chemical Name or Material tert-Butyl N-{2-[2-amino-4-(trifluoromethyl)anilino]ethyl}carbamate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'