Learn More
tert-Butyl N-(2-cyanoethyl)carbamate, 97%, Thermo Scientific™
£70.20 - £405.00
Chemical Identifiers
CAS | 53588-95-7 |
---|---|
Molecular Formula | C8H14N2O2 |
Molecular Weight (g/mol) | 170.21 |
MDL Number | MFCD02180880 |
InChI Key | NORLFIHQJFOIGS-UHFFFAOYSA-N |
Synonym | tert-butyl n-2-cyanoethyl carbamate, tert-butyl 2-cyanoethyl carbamate, n-boc-2-cyano-ethylamine, 2-cyano-ethyl-carbamic acid tert-butyl ester, t-butyl n-2-cyanoethyl carbamate, tert-butoxy-n-2-cyanoethyl carboxamide, 1,1-dimethylethyl 2-cyanoethyl carbamate, 3-tert-butoxycarbonyl amino propanenitrile, carbamicacid,n-2-cyanoethyl-,1,1-dimethylethylester |
PubChem CID | 2735664 |
IUPAC Name | tert-butyl N-(2-cyanoethyl)carbamate |
SMILES | CC(C)(C)OC(=O)NCCC#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10079481
|
Thermo Scientific Maybridge
SPB08399DA |
1 g | Amber Glass Bottle |
£70.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10160992
|
Thermo Scientific Maybridge
SPB08399EA |
10 g | Amber Glass Bottle |
£405.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
53588-95-7 | |
170.21 | |
NORLFIHQJFOIGS-UHFFFAOYSA-N | |
2735664 | |
CC(C)(C)OC(=O)NCCC#N |
C8H14N2O2 | |
MFCD02180880 | |
tert-butyl n-2-cyanoethyl carbamate, tert-butyl 2-cyanoethyl carbamate, n-boc-2-cyano-ethylamine, 2-cyano-ethyl-carbamic acid tert-butyl ester, t-butyl n-2-cyanoethyl carbamate, tert-butoxy-n-2-cyanoethyl carboxamide, 1,1-dimethylethyl 2-cyanoethyl carbamate, 3-tert-butoxycarbonyl amino propanenitrile, carbamicacid,n-2-cyanoethyl-,1,1-dimethylethylester | |
tert-butyl N-(2-cyanoethyl)carbamate |
Specifications
53588-95-7 | |
97% | |
(CH3)3COC(O)NH(CH2)2CN | |
tert-butyl n-2-cyanoethyl carbamate, tert-butyl 2-cyanoethyl carbamate, n-boc-2-cyano-ethylamine, 2-cyano-ethyl-carbamic acid tert-butyl ester, t-butyl n-2-cyanoethyl carbamate, tert-butoxy-n-2-cyanoethyl carboxamide, 1,1-dimethylethyl 2-cyanoethyl carbamate, 3-tert-butoxycarbonyl amino propanenitrile, carbamicacid,n-2-cyanoethyl-,1,1-dimethylethylester | |
CC(C)(C)OC(=O)NCCC#N | |
170.21 | |
170.21 | |
Tert-butyl N-(2-cyanoethyl)carbamate |
97.0 | |
C8H14N2O2 | |
MFCD02180880 | |
NORLFIHQJFOIGS-UHFFFAOYSA-N | |
tert-butyl N-(2-cyanoethyl)carbamate | |
2735664 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
Your input is important to us. Please complete this form to provide feedback related to the content on this product.