missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl N-[4-(aminomethyl)phenyl]carbamate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 95% |
Quantity | 10 g |
Formula Weight | 222.29 |
Percent Purity | 97% |
Chemical Name or Material | tert-Butyl N-[4-(aminomethyl)phenyl]carbamate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'