missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl nitrite, 90%, pure, Thermo Scientific Chemicals
£72.56 - £1114.55
Chemical Identifiers
CAS | 540-80-7 |
---|---|
InChI Key | IOGXOCVLYRDXLW-UHFFFAOYSA-N |
Synonym | t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono |
PubChem CID | 10906 |
IUPAC Name | tert-butyl nitrite |
SMILES | CC(C)(C)ON=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10702071
|
Thermo Scientific Acros
216221000 |
100 g | Glass bottle |
£72.56
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10452791
|
Thermo Scientific Acros
216225000 |
500 g | Glass bottle |
£253.26
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11985161
|
Thermo Scientific Acros
216220025 |
2.5 kg | Plastic drum |
£1114.55
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
540-80-7 | |
t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono | |
tert-butyl nitrite |
IOGXOCVLYRDXLW-UHFFFAOYSA-N | |
10906 | |
CC(C)(C)ON=O |
Especificaciones
540-80-7 | |
0.8600g/mL | |
−10°C | |
88% min. (GC) | |
(CH3)3CNO2 | |
01,382 | |
15,1586 | |
t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono | |
IOGXOCVLYRDXLW-UHFFFAOYSA-N | |
tert-butyl nitrite | |
10906 | |
90% | |
Liquid |
Yellow | |
61.0°C to 63.0°C | |
Authentic | |
C4H9NO2 | |
1.3670 to 1.3700 | |
07,48 | |
0.86 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,chloroform and carbon,disulfide,practically insoluble in glycerol | |
CC(C)(C)ON=O | |
103.12 | |
103.12 | |
Pure | |
tert-Butyl nitrite |
Seguridad y manipulación
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-757-
Nombre del producto
Palabra de advertencia
Clasificación
Indicaciones de peligro
Consejos de prudencia
- tert-Butyl nitrite
Palabra de advertencia
- Danger
Clasificación
- Acute toxicity Category 4
- Flammable liquid Category 2
Indicaciones de peligro
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Consejos de prudencia
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only