missing translation for 'onlineSavingsMsg'
Learn More
tert-Butylchlorodimethylsilane, 98%, Thermo Scientific Chemicals Product Code.: 10712041

tert-Butylchlorodimethylsilane, 98%, Thermo Scientific Chemicals

Product Code. 10712041
100 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10712041

Brand: Thermo Scientific Acros 183931000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18162-48-6
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.73
MDL Number MFCD00000501
InChI Key BCNZYOJHNLTNEZ-UHFFFAOYSA-N
Synonym tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride
PubChem CID 28928
ChEBI CHEBI:85071
IUPAC Name tert-butyl-chloro-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)Cl

Specifications

Melting Point 85.0°C to 91.0°C
Color Colorless to White
Density 0.8700g/mL
Boiling Point 125.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CSi(CH3)2Cl
Quantity 100 g
UN Number 2925
Fieser 04,176; 05,74; 06,78; 08,59; 09,77; 10,62; 13,87; 17,5
Merck Index 15, 1567
Specific Gravity 0.87
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohols, organic acids and amines, soluble in ethers and hydrocarbons
Formula Weight 150.73
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material tert-Butylchlorodimethylsilane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467621 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number183930250,183930050,183935000,183931000
Lot NumberA0464389 Certificate TypeCertificate of Analysis Date08/07/2024 Catalog Number183930250,183930050,183935000,183931000
Lot NumberA0456886 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog Number183930250,183930050,183935000,183931000
Lot NumberA0454038 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number183930250,183930050,183935000,183931000
Lot NumberA0392431 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number183930250,183930050,183935000,183931000
compliance-icons compliance-icons
Product Identifier
  • tert-Butylchlorodimethylsilane
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H228-Flammable solid.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butylchlorodimethylsilane, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.