missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylchlorodiphenylsilane, 98%

Product Code. 10467560 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10467560

Brand: Thermo Scientific Acros 187500500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58479-61-1 | C16H19ClSi | 274.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58479-61-1
Molecular Formula C16H19ClSi
Molecular Weight (g/mol) 274.86
MDL Number MFCD00000497
InChI Key MHYGQXWCZAYSLJ-UHFFFAOYSA-N
Synonym tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e
PubChem CID 94078
IUPAC Name tert-butyl-chloro-diphenylsilane
SMILES CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl

Specifications

Color Brown or Colorless to Yellow
Density 1.0600g/mL
Boiling Point 135°C (0.7 mmHg)
Flash Point 149°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CSi(C6H5)2Cl
Refractive Index 1.5665 to 1.5685
Quantity 50 g
Merck Index 15, 1568
Specific Gravity 1.06
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in non-protic solvents,e.g. aliphatic,or aromatic (chlorinated) hydrocarbons
Viscosity 16 mPa.s (25°C)
Formula Weight 274.86
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material tert-Butylchlorodiphenylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butylchlorodiphenylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.