missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldiphenylchlorosilane, 97%, Thermo Scientific Chemicals

£26.93 - £234.17

Chemical Identifiers

CAS 58479-61-1
Molecular Formula C16H19ClSi
Molecular Weight (g/mol) 274.863
MDL Number MFCD00000497
InChI Key MHYGQXWCZAYSLJ-UHFFFAOYSA-N
Synonym tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e
PubChem CID 94078
IUPAC Name tert-butyl-chloro-diphenylsilane
SMILES CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11490653
View Documents
Thermo Scientific Alfa Aesar
A12721.09
10 g
£26.93
10g
Please to purchase this item. Need a web account? Register with us today!
11400663
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12721.18
50 g
£73.06
50g
Please to purchase this item. Need a web account? Register with us today!
11410663
View Documents
Thermo Scientific Alfa Aesar
A12721.30
250 g
£234.17
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

tert-Butyldiphenylchlorosilane acts as a silylating reagent used to protect alcohols and in the preparation of silyl ethers. It plays a major role as a raw material and a precursor in organic synthesis and pharmaceuticals. It is also used for the synthesis of interphenylene phenyloxazoles which can be used for the treatment of circulatory disorders, angina and stroke.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyldiphenylchlorosilane acts as a silylating reagent used to protect alcohols and in the preparation of silyl ethers. It plays a major role as a raw material and a precursor in organic synthesis and pharmaceuticals. It is also used for the synthesis of interphenylene phenyloxazoles which can be used for the treatment of circulatory disorders, angina and stroke.

Solubility
Miscible with chloroform and ethyl acetate. Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, water and bases.
Specifications

Chemical Identifiers

58479-61-1
274.863
MHYGQXWCZAYSLJ-UHFFFAOYSA-N
94078
CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
C16H19ClSi
MFCD00000497
tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e
tert-butyl-chloro-diphenylsilane

Specifications

90°C to 92°C (0.01mmHg)
C16H19ClSi
UN2987
112°C (233°F)
Miscible with chloroform and ethyl acetate. Immiscible with water.
CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
274.863
274.87
1.568
1.072
58479-61-1
MFCD00000497
644023
tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e
MHYGQXWCZAYSLJ-UHFFFAOYSA-N
tert-butyl-chloro-diphenylsilane
94078
97%
Moisture sensitive
tert-Butyldiphenylchlorosilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 261-282-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only