missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals

Product Code. 10487170
Click to view available options
Quantity:
5 g
250 g
500 g
2 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1948-33-0
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00002344
InChI Key BGNXCDMCOKJUMV-UHFFFAOYSA-N
Synonym tert-butylhydroquinone, tbhq, 2-tert-butylhydroquinone, t-butylhydroquinone, mtbhq, t-butyl hydroquinone, 2-tert-butyl-1,4-benzenediol, 2-t-butylhydroquinone, mono-tert-butylhydroquinone, sustane
PubChem CID 16043
ChEBI CHEBI:78886
IUPAC Name 2-tert-butylbenzene-1,4-diol
SMILES CC(C)(C)C1=C(C=CC(=C1)O)O

Specifications

Melting Point 125.0°C to 130.0°C
Color Black-Tan to White
Boiling Point 295.0°C
Flash Point 171°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula (CH3)3CC6H3(OH)2
Quantity 2 kg
Beilstein 06, IV, 6013
Solubility Information Solubility in water: 1g/100mL (25 c). Other solubilities: soluble in acetone (190/100g at 25 c),,soluble in ethanol (120/100g at 25 c),,soluble in ethyl acetate (100g/100g at 25 c),,soluble in dmso,diethyl ether;,insoluble in benzene,toluene
Formula Weight 166.22
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material tert-Butylhydroquinone, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butylhydroquinone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'