Learn More
tert-Butyllithium, 1.9M solution in pentane, AcroSeal™, Thermo Scientific Chemicals
£60.80 - £214.07
Chemical Identifiers
CAS | 594-19-4, 109-66-0 |
---|---|
Molecular Formula | C4H9Li |
Molecular Weight (g/mol) | 64.06 |
MDL Number | MFCD00008795 |
InChI Key | BKDLGMUIXWPYGD-UHFFFAOYSA-N |
Synonym | tert-butyllithium, t-butyllithium, tbuli, lithium, 1,1-dimethylethyl, t-buli, lithium 2-methylpropane, tert-butyllithium solution, tert butyllithium, t-butyllithiurn |
PubChem CID | 638178 |
SMILES | [Li]C(C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10792135
|
Thermo Scientific Acros
396541000 |
100 mL | AcroSeal™ Glass bottle |
£60.80
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10105433
|
Thermo Scientific Acros
396548000 |
800 mL | AcroSeal™ Glass bottle |
£214.07
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
594-19-4, 109-66-0 | |
64.06 | |
BKDLGMUIXWPYGD-UHFFFAOYSA-N | |
638178 |
C4H9Li | |
MFCD00008795 | |
tert-butyllithium, t-butyllithium, tbuli, lithium, 1,1-dimethylethyl, t-buli, lithium 2-methylpropane, tert-butyllithium solution, tert butyllithium, t-butyllithiurn | |
[Li]C(C)(C)C |
Specifications
594-19-4, 109-66-0 | |
0.6900g/mL | |
−56°C | |
(CH3)3CLi | |
0.69 | |
BKDLGMUIXWPYGD-UHFFFAOYSA-N | |
lithium;2-methylpropane | |
638178 | |
17 to 19% (as BuLi) | |
tert-Butyllithium |
Colorless to Yellow | |
36.0°C to 38.0°C | |
C4H9Li | |
MFCD00008795 | |
tert-butyllithium, t-butyllithium, tbuli, lithium, 1,1-dimethylethyl, t-buli, lithium 2-methylpropane, tert-butyllithium solution, tert butyllithium, t-butyllithiurn | |
[Li]C(C)(C)C | |
64.06 | |
64.04 | |
Solution |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Extremely flammable liquid and vapor
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from any possible contact with wa
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-831-5
- tert-Butyllithium, 1.9M solution in pentane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 1
- Pyrophoric liquid Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H224-Extremely flammable liquid and vapour.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P223-Do not allow contact with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.