Learn More
tert-Butylmagnesium chloride, 1.7M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
£69.55 - £338.69
Chemical Identifiers
CAS | 677-22-5, 60-29-7 |
---|---|
Molecular Formula | C4H9ClMg |
Molecular Weight (g/mol) | 116.87 |
MDL Number | MFCD00000465 |
InChI Key | ZDRJSYVHDMFHSC-UHFFFAOYSA-M |
Synonym | tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgcl |
PubChem CID | 2724198 |
IUPAC Name | magnesium;2-methylpropane;chloride |
SMILES | CC(C)(C)[Mg]Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10688123
|
Thermo Scientific Acros
394301000 |
100 mL | AcroSeal™ Glass Bottle in Tin Can |
£69.55
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10570744
|
Thermo Scientific Acros
394308000 |
800 mL | AcroSeal™ Glass Bottle |
£338.69
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
677-22-5, 60-29-7 | |
116.87 | |
ZDRJSYVHDMFHSC-UHFFFAOYSA-M | |
2724198 | |
CC(C)(C)[Mg]Cl |
C4H9ClMg | |
MFCD00000465 | |
tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgcl | |
magnesium;2-methylpropane;chloride |
Specifications
677-22-5, 60-29-7 | |
C4H9ClMg | |
MFCD00000465 | |
ZDRJSYVHDMFHSC-UHFFFAOYSA-M | |
magnesium;2-methylpropane;chloride | |
2724198 | |
tert-Butylmagnesium chloride |
0.8g/mL | |
(CH3)3CMgCl | |
tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgcl | |
CC(C)(C)[Mg]Cl | |
116.87 | |
116.87 |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if swallowed.
In contact with water releases flammable gases which may ignite spontaneously.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Reacts vio
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- tert-Butylmagnesium chloride, solution in diethylether
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H224-Extremely flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only