missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylphosphonic acid, 98%

Product Code. 10638221 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10638221

Brand: Thermo Scientific Acros 321520010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4923-84-6 | C4H9O3P | 136.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4923-84-6
Molecular Formula C4H9O3P
Molecular Weight (g/mol) 136.09
MDL Number MFCD00012296
InChI Key OGDSVONAYZTTDA-UHFFFAOYSA-L
Synonym phosphonic acid, 1,1-dimethylethyl, acmc-20ajka, t-butylphosphonic acid, tert-butylphosphonic acid, 2-methylprop-2-ylphosphonic acid, phosphonicacid, 1,1-dimethylethyl
PubChem CID 312546
IUPAC Name tert-butylphosphonic acid
SMILES CC(C)(C)P([O-])([O-])=O

Specifications

Melting Point 188.0°C to 192.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)3CP(O)(OH)2
Quantity 1 g
Formula Weight 138.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material tert-Butylphosphonic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.