missing translation for 'onlineSavingsMsg'
Learn More
tert-Pentylamine, 98%, Thermo Scientific Chemicals Product Code.: 11470498

tert-Pentylamine, 98%, Thermo Scientific Chemicals

Product Code. 11470498
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11470498

Brand: Thermo Scientific Alfa Aesar B24639.14

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-39-8
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00008056
InChI Key GELMWIVBBPAMIO-UHFFFAOYSA-N
Synonym tert-amylamine, tert-pentylamine, 2-butanamine, 2-methyl, t-amylamine, 1,1-dimethylpropylamine, 2-methyl-2-butylamine, 2-amino-2-methylbutane, 2-methyl-butan-2-amine, 1,1-dimethyl-1-propylamine, t-pentylamine
PubChem CID 68986
ChEBI CHEBI:84236
IUPAC Name 2-methylbutan-2-amine
SMILES CCC(C)(C)N

Specifications

Density 0.746
Boiling Point 76°C to 78°C
Flash Point −1°C (30°F)
Odor Amine-like
Refractive Index 1.3995
Quantity 25 g
UN Number UN1106
Sensitivity Air Sensitive
Formula Weight 87.17
Percent Purity 98%
Chemical Name or Material tert-Pentylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Pentylamine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.