missing translation for 'onlineSavingsMsg'
Learn More

tert-Pentylamine, 98%

Product Code. p-7027687
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11470498

Brand: Thermo Scientific Alfa Aesar B24639.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 594-39-8 | C5H13N | 87.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-39-8
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00008056
InChI Key GELMWIVBBPAMIO-UHFFFAOYSA-N
Synonym tert-amylamine, tert-pentylamine, 2-butanamine, 2-methyl, t-amylamine, 1,1-dimethylpropylamine, 2-methyl-2-butylamine, 2-amino-2-methylbutane, 2-methyl-butan-2-amine, 1,1-dimethyl-1-propylamine, t-pentylamine
PubChem CID 68986
ChEBI CHEBI:84236
IUPAC Name 2-methylbutan-2-amine
SMILES CCC(C)(C)N

Specifications

Density 0.746
Boiling Point 76°C to 78°C
Flash Point −1°C (30°F)
Odor Amine-like
Refractive Index 1.3995
Quantity 25 g
UN Number UN1106
Sensitivity Air Sensitive
Formula Weight 87.17
Percent Purity 98%
Chemical Name or Material tert-Pentylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Pentylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : tert-Amylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.