missing translation for 'onlineSavingsMsg'
Learn More

TES, 0.2M buffer soln., pH 8.0, Thermo Scientific™

£43.20 - £64.39

Chemical Identifiers

CAS 70331-82-7
Molecular Formula C6H14NNaO6S
Molecular Weight (g/mol) 251.23
MDL Number MFCD00065482
InChI Key GUXMFAHOYNRHBI-UHFFFAOYSA-M
PubChem CID 23702152
SMILES [Na+].OCC(CO)(CO)NCCS([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15424379
View Documents
Thermo Scientific Alfa Aesar
J61889.AE
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15434379
View Documents
Thermo Scientific Alfa Aesar
J61889.AK
250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
TES, 0.2M buffer soln., is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9, and is also useful for studying succinate oxidation. It is also widely used as a buffer in biological applications.

Solubility
Soluble in water (350 g/L) at 20°C, and methanol(very faint turbidity).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

70331-82-7
251.23
GUXMFAHOYNRHBI-UHFFFAOYSA-M
[Na+].OCC(CO)(CO)NCCS([O-])(=O)=O
C6H14NNaO6S
MFCD00065482
23702152

Specifications

70331-82-7
8.0
C6H14NNaO6S
Soluble in water (350g/L) at 20°C,and methanol(very faint turbidity).
[Na+].OCC(CO)(CO)NCCS([O-])(=O)=O
251.23
TES buffer soln.
0.2 M
Liquid
MFCD00065482
GUXMFAHOYNRHBI-UHFFFAOYSA-M
sodium;2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonate
23702152
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications