missing translation for 'onlineSavingsMsg'
Learn More

TES, 99%

Product Code. 11464847
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11464847

Brand: Thermo Scientific Alfa Aesar B21819.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7365-44-8 | C6H15NO6S | 229.247 g/mol

TES plays an important role in production of pH buffers for a biological system. It is used to make zwitterionic buffer solutions in the pH range 6.8-8.2. It is used in molecular biology, diagnosis, cell culture, pharmacy, agrochemical and pharmaceuticals. It is also used in the study of succinate oxidation and one of the components of test yolk buffer medium used in refrigeration of semen and transport.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TES plays an important role in production of pH buffers for a biological system. It is used to make zwitterionic buffer solutions in the pH range 6.8-8.2. It is used in molecular biology, diagnosis, cell culture, pharmacy, agrochemical and pharmaceuticals. It is also used in the study of succinate oxidation and one of the components of test yolk buffer medium used in refrigeration of semen and transport.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-44-8
Molecular Formula C6H15NO6S
Molecular Weight (g/mol) 229.247
MDL Number MFCD00007532
InChI Key JOCBASBOOFNAJA-UHFFFAOYSA-N
Synonym tes, n-tris hydroxymethyl methyl-2-aminoethanesulfonic acid, tes buffer, tes free acid, ethanesulfonic acid, 2-2-hydroxy-1,1-bis hydroxymethyl ethyl amino, 2-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino ethanesulfonic acid, 2-2-hydroxy-1,1-dihydroxymethyl-ethylamino-ethanesulfonic acid, tris hydroxymethyl methyl-2-aminomethane sulfonic acid, 2-tris hydroxymethyl methylamino ethane-1-sulphonic acid, n-tri hydroxymethyl methyltaurine
PubChem CID 81831
ChEBI CHEBI:44356
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid
SMILES C(CS(=O)(=O)O)NC(CO)(CO)CO

Specifications

Melting Point ∼225°C (decomposition)
Odor Odorless
Linear Formula (HOCH2)3CNHCH2CH2SO3H
Quantity 50 g
Beilstein 1957061
Merck Index 14,9176
Solubility Information Soluble in water.
Formula Weight 229.25
Percent Purity 99%
Chemical Name or Material TES

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.