Learn More
Palintest™ Test Kit Chlorine DPD 1 Refill
Reliable, well-established, cost-effective test procedure for free, total, and combined chlorine, bromine, and ozone.
Brand:  Palintest™ AP011
Product Code. 13111461
  • £69.90 / 250 tests
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Test procedure for free, total, and combined chlorine, bromine, and ozone
  • Measures free available chlorine with result in mg/L chlorine
  • Widely used testing methods for free and total chlorine 
Specifications
Specifications
250/Pk.
250/Pk.