Learn More
IKA™ Test Tube Attachment for IKA Platform Shaker
Attachment with customized array for holding reagent glasses on a platform shaker.
£30.00 - £74.00
Specifications
Product Type | Test Tube Adapter Attachment |
---|---|
Description | Test tube adapter attachment |
For Use With (Equipment) | Universal attachment, shaker |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
10490243
|
IKA™
L001830 |
Test Tube Adapter Attachment |
£30.00
Each |
Estimated Shipment: 23-10-2024
Log in to see stock. |
|||||
10728971
|
IKA™
L001840 |
Test Tube Adapter Attachment |
£74.00
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
|||||
10130921
|
IKA™
L001850 |
Test Tube Adapter Attachment |
£61.00
Each |
Estimated Shipment: 23-10-2024
Log in to see stock. |
|||||
Description
Attachment with customized array for holding reagent glasses on a platform shaker.
- Requires VG 3.3 universal attachment
- Facilitates multi-tube vortexing
Specifications
Test Tube Adapter Attachment | |
Test tube adapter attachment | |
Universal attachment, shaker |