Learn More
IKA™ Test Tube Attachment for IKA Vortex Shaker
Attachment with customized array for holding reagent glasses on a vortexed.
Brand:  IKA™ 0003344200
Code : 50
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10414522
  • £39.00 / Each
Estimated Shipment: 01-10-2024
to see stock.
Add to basket
Description
Description
  • Requires VG 3.3 universal attachment
  • Facilitates multi-tube vortexing
Specifications
Specifications
8 x 20 mm Reagent Glasses
Test Tube Adapter Attachment
Model VG 3.35
8
20 mm
Test tube adapter attachment
VG 3.3 Universal Attachment, Vortexer
VG 3.3 Universal Attachment