Learn More
IKA™ Test Tube Attachment for IKA Vortex Shaker
Attachment with customized array for holding reagent glasses on a vortexed.
Brand:  IKA™ 0003344100
Code : 50
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10656263
  • £39.00 / Each
Estimated Shipment: 29-10-2024
to see stock.
Add to basket
Description
Description
  • Requires VG 3.3 universal attachment
  • Facilitates multi-tube vortexing
Specifications
Specifications
8 x 16 mm Reagent Glasses
Test Tube Adapter Attachment
Model VG 3.34
8
16 mm
Test tube adapter attachment
VG 3.3 Universal Attachment, Vortexer,
VG 3.3 Universal Attachment