Learn More
OHAUS™ Test Tube Holder
Hands free mixing designed to fit on the Heavy Duty Vortex Mixer. Easily attached to the top of mixer and is magnetically secured.
Brand:  OHAUS™ 30400222
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15742639
  • £35.30 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Use with Heavy Duty Vortex Mixer
Specifications
Specifications
18 x 19 to 21 mm Test Tubes
Foam Insert
Foam Insert For (18) 19-21 mm Tubes
1 Ea.
19 to 21 mm
Foam
Vortex Mixer