Learn More
Labconco™ Test Tube/Microcentrifuge Tube Rack: FreeZone™ Stoppering or Bulk Tray Dryer Accessory
Rectangular rack for 12-13 mm test tubes and microcentrifuge tubes.
Brand:  Labconco™ 7370001
Product Code. 17371773
  • £329.00 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Rectangular Test Tube/Microcentrifuge Rack
  • Holds up to 70 12-13 mm tubes
  • Made of type 304 stainless steel
  • Includes bottom & adapter plate for (70) microcentrifuge tubes
  • Ideal for use with FreeZone™ Tray Dryer, Bulk Tray Dryer, Stoppering Tray Dryer
  • Dimensions: 7.4 in. w x 11.5 in. d x 2.5 in. h
Specifications
Specifications
Stainless Steel (304)
Stainless steel rack and 1 stainless steel microcentrifuge tube adapter plate
2.5 in.
3.5 lb.
FreeZone™ Tray Dryer, Bulk Tray Dryer, or Stoppering Tray Dryer
7.4 in.
11.5 in.
2 lb.