Learn More
Memmert™ Test Tube Rack
Test Tube Rack designed for use with a Memmert™ Water Bath.
Brand: Memmert™ WTB50B50662
This item is not returnable.
View return policy
Description
- Water Bath Accessory
Specifications
WTB50 Water Baths | |
75 Test Tubes (3 Rows) | |
18 mm |
Test Tube Rack | |
Stainless Steel |