Learn More
Memmert™ Test Tube Rack
Test Tube Rack designed for use with a Memmert™ Water Bath.
Brand:  Memmert™ WTB50B50658
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 17562265
  • £71.45 / Each
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
Description
Description
  • Water Bath Accessory
Specifications
Specifications
WTB50 Water Baths
58 Test Tubes (2 Rows)
14.5 mm
Test Tube Rack
Stainless Steel