Learn More
Memmert™ Test Tube Racks with 24 Holes
Test tube rack with 24 holes and 2-row rack
Brand: Memmert™ WTB11B48828
Additional Details : Weight : 0.01100kg
Description
- Test tube rack
- 24 holes 2-row rack
Specifications
24 holes, 2-row rack | |
Test Tube Rack | |
18 mm |
WTB11 | |
Transverses for sizes 15 and 24 |