Learn More
Memmert™ Test Tube Racks with 48 Holes
Test tube rack with 48 holes and 4-row rack
Brand:  Memmert™ WTB15B48837
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 17110335
  • £107.00 / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Description
Description
  • Test tube rack
  • 48 holes 4-row rack
  • Diameter is 18 mm
Specifications
Specifications
48 holes, 4-row rack
Test Tube Rack
18 mm
WTB15
Longitudinals for sizes 15 and 24