missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Test Tube Racks for 5 L Baths
Test tube racks for use with Fisherbrand™ 5 L Digital Heated Circulating Baths.
Brand: Fisherbrand™
Additional Details : Weight : 0.30000kg
Description
- Conveniently stores test tubes being used with Fisherbrand™ 5 L Digital Heated Circulating Baths
Specifications
Test Tubes | |
16 x 16 to 19 mm Tubes |